KingFisher™ Presto Purification System
Fournisseur: Thermo Fisher Scientific
5400830
FINN5400830EA 50700 EUR InStock
FINN5400830
KingFisher™ Presto Purification System
Systèmes de purification des acides nucléiques
Use the Thermo Scientific KingFisher Presto as part of an automated purification system with a liquid handler for plate filling in high-throughput laboratories.
  • Volume range of 50 to 5000 μl
  • Minimised size for easy fitting to automation systems
  • Heating capability for application requiring elevated temperatures
  • Easy connection to several liquid handling instruments
  • Flexible system for wide variety of applications
Utilising proven KingFisher magnetic particle-based technology, the KingFisher Presto provides high-quality yields of target nucleic acids and proteins for biopharma, biotech and research.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité