Ready DNA Ultra 2.0 prefilled plates, for KingFisher Flex instruments, KingFisher™
Fournisseur: Thermo Fisher Scientific
A36586
FINNA36586EA 812 EUR InStock
FINNA36586
Ready DNA Ultra 2.0 prefilled plates, for KingFisher Flex instruments, KingFisher™
Systèmes de purification des acides nucléiques
KingFisher Ready DNA Ultra 2.0 Prefilled plates are automation-ready plates prefilled with MagMAX DNA Multi-Sample Ultra 2.0 reagents suitably configured for KingFisher Flex purification systems. KingFisher Ready DNA Ultra 2.0 prefilled plates enable isolation of genomic DNA from 50 to 400 µl of multiple sample types such as whole blood, saliva, buffy coat, buccal swabs, and more. The purified gDNA is ideal for a variety of downstream molecular biology applications.
  • Maximise convenience with MagMAX DNA Multi-Sample Ultra 2.0 isolation chemistry prefilled into KingFisher SBS microtiter plates
  • Increase ease of use - process 96 samples on instrument without the hassle of reagent setup
  • Designed to reduce user error
  • Helps to reduce hands-on time
  • Formatted specifically for KingFisher Flex instruments
Contents: 1 complete set of KingFisher Ready DNA Ultra 2.0 Prefilled plates, 4,5 ml proteinase K solution, 4,5 ml enhancer solution, 1 KingFisher Flex 96-tip comb.
Attention: Store all contents in an upright position and at room temperature.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité