CO₂ incubators, Forma™ Steri-Cycle™
Fournisseur: FORMA (TF-GROUP)
381 371TF
THBR381EA 0 EUR InStock
THBR381 FORM371TF
CO₂ incubators, Forma™ Steri-Cycle™
Incubateurs CO₂ Incubators
CO₂ incubators with HEPA filtration system to provide continuous protection against airborne contaminants with high-temperature sterilisation cycle that simplifies routine cleaning practices. Made of stainless steel interior with coved corners for easy cleaning, units have stainless steel shelves and supports are designed to support a full product load.
  • Precise temperature control and choice of TC (thermal conductivity) or IR (infrared) sensor
  • Convenient high-temperature decontamination cycle to destroy all contaminants
  • VOC filtration system removes volatile organic vapours which could pose a risk to sensitive cultures
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité