Permanent Deep Freeze Cover Up Labels for Thermal Transfer Printers
Fournisseur: GA
BOC-127C1-1WH BOC-127C3-2WH BOC-152C1-1WH BOC-152C3-6WH BOC-243C1-1WH BOC-243C3-2WH BOC-27C1-0.5WH BOC-27C3-1WH BOC-28C1-1WH BOC-28C3-2WH BOC-507C1-1WH BOC-507C3-2WH BOC508C1-0.5WH BOC-508C3-1WH BOC521C10.15WH BOC521C3-0.3WH BOC529C1-0.5WH BOC-529C3-1WH BOC530C1-0.5BL BOC530C1-0.5LA BOC530C1-0.5WH BOC-530C3-2BL BOC-530C3-2LA BOC-530C3-2WH BOC-5C1-1WH BOC-5C3-2WH BOC-62C1-0.5WH BOC-62C3-1WH BOC-7C1-1WH BOC-7C3-5WH BOC-86C1-1WH BOC-86C3-2WH BOC-97C1-1WH BOC-97C3-2WH
GAINBOC-127C1-1WHEA 175 EUR InStock
GAINBOC-127C1-1WH GAINBOC-127C3-2WH GAINBOC-152C1-1WH GAINBOC-152C3-6WH GAINBOC-243C1-1WH GAINBOC-243C3-2WH GAINBOC-27C1-0.5WH GAINBOC-27C3-1WH GAINBOC-28C1-1WH GAINBOC-28C3-2WH GAINBOC-507C1-1WH GAINBOC-507C3-2WH GAINBOC508C1-0.5WH GAINBOC-508C3-1WH GAINBOC521C10.15WH GAINBOC521C3-0.3WH GAINBOC529C1-0.5WH GAINBOC-529C3-1WH GAINBOC530C1-0.5BL GAINBOC530C1-0.5LA GAINBOC530C1-0.5WH GAINBOC-530C3-2BL GAINBOC-530C3-2LA GAINBOC-530C3-2WH GAINBOC-5C1-1WH GAINBOC-5C3-2WH GAINBOC-62C1-0.5WH GAINBOC-62C3-1WH GAINBOC-7C1-1WH GAINBOC-7C3-5WH GAINBOC-86C1-1WH GAINBOC-86C3-2WH GAINBOC-97C1-1WH GAINBOC-97C3-2WH
Permanent Deep Freeze Cover Up Labels for Thermal Transfer Printers
Étiquettes
These freezer grade blackout labels are designed to conceal any already existing information or labels. Can be used to re-label cryo vials, microtubes, and freezer boxes stored in ultra-low temperature freezers (−80 °C) or transported in dry ice.
  • Temperature range: −80 to +120 °C
  • Unique opaque design allows the labels to obscure all pre-existing information
  • Compatible with all major thermal transfer printer brands
Deep-freeze permanent blackout labels are designed to conceal any already existing information or labels. Ideal for re-labelling cryo vials, microtubes, and freezer boxes, to be stored in ultra-low temperature freezers (−80 °C) or to be transported in dry ice. These cover-up labels will completely obscure the existing label and block out the information underneath.
Printable with all major brands of thermal transfer printers, these versatile labels can withstand a wide temperature range, and are also suitable for high-temperature applications, up to 120 °C.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité