Lab-TAG™ Cryogenic Labels, White
Fournisseur: GA INTERNATIONAL INC.
SPJTTA4NPC31WH
GAINSPJTTA4NPC31WHEA 40.3 EUR InStock
GAINSPJTTA4NPC31WH
Lab-TAG™ Cryogenic Labels, White
Étiquettes
Cryogenic labels for identifying small diameter cryo vials, tubes, plates, cardboard freezer boxes, polypropylene cryo-boxes, and plastic bags.
  • Thermal Transfer Print Technology
These cryo labels feature our new patent-pending SimPEEL™ technology, making peeling them off their liner quick and easy. Their innovative adhesive-free tab design also makes the label’s application on their destined container hassle-free, as they won’t stick to your hands or gloves, eliminating in this fashion adhesive contamination. Ideal for routine, high-volume container identification in hospitals, clinics, blood collection centers, biotechnology companies, biorepositories, and others. They are designed for long-term cryo storage under cryogenic laboratory conditions, including liquid nitrogen −196 °C (−321 °F), freezers −80 °C (−112 °F), and for dry ice transport. Labels can be printed with all major brands of thermal-transfer printers.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité