Transparent Tamper-Evident Cryogenic Tape for Frozen Containers
Fournisseur: GA INTERNATIONAL INC.
TEQ-22C3-100
GAINTEQ-22C3-100EA 90.5 EUR InStock
GAINTEQ-22C3-100
Transparent Tamper-Evident Cryogenic Tape for Frozen Containers
Étiquettes
Transparent tamper-evident tape for already frozen containers and boxes, ideal for long-term storage in cryogenic and deep-freeze conditions. Suitable for immersion and thawing in a water baths, the tape will tear upon any attempt to remove it, indicating any attempt to open the labeled container.
  • Temperature range: –196 to +121 °C (–321 to +250 °F)
  • Clear tamper-evident design indicates any attempt to open the container
  • Writable with permanent cryogenic markers
  • Ideal for already frozen surfaces (as low as –80 °C) without thawing first
Transparent tape comes in a roll format to identify already frozen plastic containers. Tapes are waterproof, suitable for immersion and thawing in a water bath, with a unique tamper-evident design. Opening of the container will tear or destroy the seal, indicating that tampering occurred and that the sample or product it contains has been compromised. Designed to be marked on with permanent cryogenic markers.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité