Affinity chromatography columns, HiScreen™ IXSelect
Fournisseur: Cytiva
17-3714-10
GEHE17-3714-10EA 1318 EUR InStock
GEHE17-3714-10
Affinity chromatography columns, HiScreen™ IXSelect
Colonnes pour chromatographie
HiScreen™ IXSelect columns are prepacked with IXSelect affinity chromatography medium. These columns are an excellent choice for method optimisation and parameter screening.
  • Easily connected in series to achieve 20 cm bed height
  • Small bed volume gives fast results and minimal sample/buffer consumption
  • Reproducible results, scalable to BioProcess columns packed with the same chromatography resin using the same linear fluid velocity
IXSelect is a high-throughput resin specifically designed for the purification of factor IX targets from recombinant feed streams.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité