HiTrap™ Capto™ Octyl Hydrophobic Interaction Chromatography Column
Fournisseur: Cytiva
17-5465-08
GEHE17-5465-08EA 247 EUR InStock
GEHE17-5465-08
HiTrap™ Capto™ Octyl Hydrophobic Interaction Chromatography Column
Colonnes pour chromatographie
Capto™ Octyl are hydrophobic interaction chromatography (HIC) resins used in the capture and intermediate stages of protein purification.
  • Improved productivity and process economy in downstream operations
  • Very high flow rates and large sample volume processing
  • Excellent chemical stability
Their combination of high capacity, narrow specification range, high flow rate and low backpressure consistently reduces process cycle times and increases productivity. The Capto™ product range comprises modern media that meet the demands of large-scale biopharmaceutical manufacturing today.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité