Human Serum AB (Off The Clot), Aseptic Assurance System
Fournisseur: GEMINI BIO-PRODUCTS
100-318-750
GEMB100-318-750EA 858 EUR InStock
GEMB100-318-750
Human Serum AB (Off The Clot), Aseptic Assurance System
Sérums
Product has minimum order quantity of 200 units. Off the clot human serum AB is collected from normal healthy male donors at FDA-licensed facilities in the United States. As a natural off the clot product, this material contains no anticoagulants/preservatives. All donor units are tested for viral markers and found to be non-reactive.
  • Supports clinical and commercial applications
  • Aseptic assurance system
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité