GemCell™ Plus Xeno-Free Human Serum, Aseptic Assurance System
Fournisseur: GEMINI BIO-PRODUCTS
100-912-750
GEMB100-912-750EA 801 EUR InStock
GEMB100-912-750
GemCell™ Plus Xeno-Free Human Serum, Aseptic Assurance System
Sérums
Product has minimum order quantity of 200 units. GemCell™ Plus Xeno-Free human serum is collected from healthy male donors of the AB serotype at FDA-licensed facilities in the United States. This material goes through additional rigorous testing and contains therapeutic grade human thrombin. All donor units are tested for viral markers and found to be non-reactive.
  • This product was developed without the use of animal material
  • Custom fill sizes in bottle and bag format offered
  • 0,1 µm sterile-filtered
  • Supports clinical and commerical applications
  • Aseptic
More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité