GemCell™ Plus Xeno-Free Human Serum, World Grade
Fournisseur: GEMINI BIO-PRODUCTS
100-916-100
GEMB100-916-100EA 691 EUR InStock
GEMB100-916-100
GemCell™ Plus Xeno-Free Human Serum, World Grade
Sérums
Product has minimum order quantity of 200 units. GemCell™ Plus Xeno-Free, world grade is collected from a maximum of 16 healthy male donors of the AB serotype. Product is defibrinated from source plasma using therapeutic grade recombinant human thrombin. Based on extensive testing and manufacturing process that is Xeno-Free, supports clinical and commercial applications.
  • This product was developed without the use of animal material
  • Custom fill sizes in bottle and bag format offered
  • 0,1 µm sterile-filtered
  • Supports clinical and commerical applications
  • Meet
More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité