The SpinOUT GT-100, GT-600 and GT-1200 columns are versatile, spin-format columns for the desalting and buffer exchange of peptide, protein and nucleic acid solutions ranging from 5l to 4ml sample volumes. Spin-OUT GT-100 is for the purification o 1 * 5 mL
Fournisseur: G Bioscienes
The SpinOUT GT-100, GT-600 and GT-1200 columns are versatile, spin-format columns for the desalting and buffer exchange of peptide, protein and nucleic acid solutions ranging from 5l to 4ml sample volumes. Spin-OUT GT-100 is for the purification o 1 * 5 mL
COMMANDE
Développer tout / Réduire tout
The SpinOUT™ GT-100, GT-600 and GT-1200 columns are versatile, spin-format columns for the desalting and buffer exchange of peptide, protein and nucleic acid solutions ranging from 5µl to 4ml sample volumes. Spin-OUT™ GT-100 is for the purification o 1 * 5 mL GENO786-868 1 * 5 mL
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou