Human HbA1c (glycosylated hemoglobin A1c) ELISA kit
Fournisseur: G Bioscienes
IT1822
GENOIT1822EA 1700 EUR InStock
GENOIT1822
Human HbA1c (glycosylated hemoglobin A1c) ELISA kit
Tests Tests ELISA
Human HbA1c (glycosylated hemoglobin A1c) ELISA kit is based on a sandwich enzyme-linked immunosorbent assay (ELISA) technology.
A 96-well plate is coated with a specific antibody against the protein of interest (HbA1c), which will bind the protein and this is detected by a biotin conjugated antibody against the protein of interest. The biotin moiety is subsequently detected, following washing, by the addition of streptavidin coupled horseradish peroxidase (HRP). The supplied TMB substrates were used to visualise HRP enzymatic reaction. TMB was catalysed by HRP to produce a blue colour product that changed into yellow after adding acidic stop solution. The density of yellow is directly proportional to the concentration of protein of interest captured on the plate.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité