GCQ CleanType® Medical Silicone Keyboard with Illumination
Fournisseur: GETT GERATETECHNIK
KG24229
GETTKG24229EA 163 EUR InStock
GETTKG24229
GCQ CleanType® Medical Silicone Keyboard with Illumination
Claviers
The CleanType® Medical keyboard is specially manufactured for medical environments. With its versatility, it can be used in almost all areas of hospitals, clinics, and medical practices.
  • IP 68 rated medical keyboard
  • Fully washable and disinfectable
  • Water-resistant silicone USB protection cap
  • IEC 60601-1-2 certified
  • For use in various areas: hospitals, offices, laboratories, production areas
The CleanType® Medical keyboard is completely waterproof, can be disinfected, and features integrated backlighting. The keyboard is a product of the GCQ series from GETT.
Certifications: CE, FCC, REACH.
Conditionnement: Keyboards are packed and delivered in environmentally friendly brown cardboard.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité