CleanType® Xtra Touch Protect Compact Layout IP 68 Silicone Keyboard with Touchpad, USB
Fournisseur: GETT GERATETECHNIK
KG30217
GETTKG30217EA 149 EUR InStock
GETTKG30217
CleanType® Xtra Touch Protect Compact Layout IP 68 Silicone Keyboard with Touchpad, USB
Claviers
Silicone keyboard with touchpad - perfect for use in medicine and industry. Wired with USB cable.
  • Precise and flat key profile due to scissor key mechanics
  • Cursor control by integrated capacitive touchpad
  • IP 68 rated: The keyboard is completely water- and dustproof which allows easy, fast and deep cleaning and disinfection
  • Keylock function: To avoid missuse during the cleaning process you can disable the buttons of the keyboard by hitting the keylock button
  • Fold-out feet to tilt the keyboard for ergonomic work
  • USB cable length: 1,8 m
  • Two year warranty
The CleanType® Touch Protect keyboard is a high-quality compact silicone keyboard that is easy to clean thanks to its water, disinfection and dust resistance, making it ideal for use in hygiene-sensitive areas such as hospitals, clinics, medical practices, laboratories or pharmaceutical companies.
USB wired version currently available in DE Layout. For other country layouts required please contact your Avantor sales representative.
Certifications: CE, FCC EN60601-1-2- Ed 4, EN61000-6-2.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité