ASPEC® 274 system
Fournisseur: GILSON LCMS
2614010
GILI2614010EA 32404 EUR InStock
GILI2614010
ASPEC® 274 system
Extracteurs Extracteurs automatiques
This ASPEC® system for positive pressure solid phase extraction (SPE) automation maximises throughput by processing up to four samples in parallel. With its small footprint and multiple probes, it is the ideal choice for reproducible and efficient sample cleanup for clinical, forensic, doping, food, and environmental testing labs.
  • Parallel processing for maximum efficiency
  • Positive pressure for reproducible extractions
  • Smart design for maximised performance
  • Intuitive error handling for trusted results
  • Powerful software control for optimised productivity
  • Versatile set-up for trace analysis
The ASPEC 274 system provides:
Four-probe Z-arm enables processing of four samples in parallel, increasing throughput and efficiency. Positive pressure extraction with unique sealing cap design increases reproducibility from sample to sample. The syringe pump delivers accurate volume dispensing and full control of sorbent drying. Large bed capacity for processing up to 144 samples unattended. The mobile racks accommodate 1, 3 and 6 ml SPE polypropylene cartridges, and enable multiple fraction elutions per sample. The syringe pump manages volumes from 1 to 10 ml with high accuracy and precision, allowing the transfer of eluted fractions in sealed injection vials or directly in the LC system with the optional GX direct injection module. The syringe pump pressure-sensing module provides error handling for high cartridge pressure or clogged liquid lines and monitors pressure on each fluid path individually. Optional jet wash rinse stations minimise carryover and cross contamination.
Commander maintenant
COMMANDE
ACCESSOIRES
PRODUITS COMPLEMENTAIRES
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité