Cryoboxes, PP, D270
Fournisseur: GLW
D270
GLWAD270EA 17.5 EUR InStock
GLWAD270
Cryoboxes, PP, D270
Boîtes Boîtes pour cryotubes
Cryoboxes are available in various heights. It is possible to customize the box according to individual storage needs.
  • Made from polypropylene
  • Autoclavable up to +121 °C and use in freezer down to –90 °C
  • The included clear lid offers a view of the contents without opening the cryobox
  • Without drainage holes in base for catching drips or store fluids (leakproof only at bottom up to 40 mm)
  • Height extension secured with exclusive clicksystem (not glued)
The box with drainage holes is ideal for rock samples and bulky items, for example. The GLW box isMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité