PCR plate centrifuge/vortex mixer, CVP-2
Fournisseur: GRANT INSTRUMENTS
CVP-2
GRANCVP-2EA 1500 EUR InStock
GRANCVP-2
PCR plate centrifuge/vortex mixer, CVP-2
Centrifugeuses Microcentrifugeuses
This unique product offers 3 devices in one: Centrifuge with vortex mixing, PCR plate centrifuge and PCR plate mixer. It allows for the simultaneous sample preparation of multiple samples at one time by providing repeating spin down and shaking of two un, semi or fully skirted PCR plates.
  • Centrifuge and vortex mixer combined for significant time saving
  • Spin and vortex mixing functions can operate independently
  • Very small footprint – saves bench space
  • Simple push-buttons and a clear 2-line LCD status display of set and actual values
  • Clear window in the lid allows the user to observe the samples
Ideal for life science applications, PCR and molecular biology preparation, thawing/mixing, master mix preparation, cell biology, cell lysis, DNA/RNA isolation and purification, pellet re-suspension, and any other method where you need tube vortexing and centrifugation.
Informations de commande: Supplied with adapters.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
SERVICES
Services: Besoin d'une installation, d'un IQOQ, d'un entretien ou d'une réparation? Contactez-nous : technical.service.be@vwr.com
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité