Digital multispeed vortex mixer, MSV-3500
Fournisseur: GRANT INSTRUMENTS
MSV-3500
GRANMSV-3500EA 522 EUR InStock
GRANMSV-3500
Digital multispeed vortex mixer, MSV-3500
Agitateurs Agitateurs Vortex
The Grant bio MSV-3500 is a versatile multi speed vortex mixer is designed for soft or intensive mixing of reagents in different size and type plastic tubes. It is designed for operation in life-science laboratories working in the fields of biochemistry, cell and molecular biology.
  • Four types of interchangeable platforms
  • Microprocessor control, LCD display
  • Acceleration time to max speed: 15 seconds
Provides high maximum speed of platform rotation efficiently mixing microvolumes (less than 5 ml) of samples.
Widely used in intensive stirring of bacterial and yeast cells, extraction of metabolites and enzymes from cells and cell cultures, and various DNA operations such as deproteinisation of DNA/protein complexes and purification of low molecular weight DNA/RNA fragments in PCR diagnostics.
Information de livraison: Platforms are sold separately or available as part of a kit (GRANMSV-3500 4P).
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité