Pressure logger, hand-held, GMH3111
Fournisseur: Greisinger
600374
GRSG600374EA 0 EUR InStock
GRSG600374
Pressure logger, hand-held, GMH3111
Enregistreurs de données
The GMH3111 pressure logger is designed with impact-resistant ABS plastic housing, membrane keyboard, transparent panel and front side IP 65. The pressure sensors are already callibrated and fully interchangeable pressure probes.
  • LCD display and auto off function
  • Automatic probe detection and setting of measuring range upon plugging in of probe
  • Direct connection to RS232 or USB  interface of a PC via electrically isolated interface converter
  • Pressure units: mbar, bar, Pa, kPa, MPa, mmHg, PSI, m, can be selected
  • Tare/Hold function, min/max memory
  • Measuring cycle: Four measurements/seconds
Certifications: ISO certified.
Information de livraison: Supplied with 9 V battery and manual.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité