Oxygen meter, handheld, GMH 3692
Fournisseur: Greisinger
605919
GRSG605919EA 0 EUR InStock
GRSG605919
Oxygen meter, handheld, GMH 3692
Electrochemistry Meters Oxygen Meters
Compact meter for measuring oxygen in air and gas mixtures either as partial pressure or as concentration in %vol. For the measuring an external sensor has to be connected to the MINI-DIN socket. Suitable for a wide range of applications, such as oxygen monitoring in breeding chambers for cell cultures, in respirators, incubators and in air conditioning systems.
  • Oxygen and temperature are displayed simultaneously
  • Automatic temperature and air pressure compensation
  • Min./Max. value memory, Hold function
  • Adjustable power-off function: from 1 to 120 min
  • Alarm function: for min. or max. limits
  • With pop-up and belt clip
Suitable for a wide range of applications, such as oxygen monitoring in breeding chambers for cell cultures, in respirators, incubators and in air conditioning systems.
Information de livraison: Supplied without sensor (needs to be ordered separately).
Commander maintenant
COMMANDE
SPÉCIFICATIONS
SERVICES
Services: Besoin d'une installation, d'un IQOQ, d'un entretien ou d'une réparation? Contactez-nous : technical.service.be@vwr.com
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité