Coronavirus and SARS-CoV-2 direct RT real-time PCR kits, GSD NovaPrime®
Fournisseur: GOLD STANDARD DIAGNOSTICS
PCOV6043 PCOV6044
GSDCPCOV6043EA 806 EUR InStock
GSDCPCOV6043 GSDCPCOV6044
Coronavirus and SARS-CoV-2 direct RT real-time PCR kits, GSD NovaPrime®
Systèmes de diagnostic clinique Réactifs et Kits de diagnostic clinique
This Real-Time RT-PCR assay is intended for the qualitative detection of SARS-CoV-2 from human nasopharyngeal swabs, eluted directly into deionised water without an additional RNA extraction process.
  • Simplify the process for COVID-19 testing
  • Meet the increasing global demand for molecular solutions
  • Maximize accessibility of RT-PCR testing to a larger number of laboratories
  • Increase test processing
  • Reduce the cost, labour, and time required to perform RT-PCR
This new workflow shortens the complete turnaround time of the test, providing results in less than 2 hours from sample to result without compromising the high accuracy of PCR testing.
Certifications: CE-IVD marked.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité