SNOOD CAP WITH HAIRNET COLOR: DARK BLUE SIZE: L DESCRIPTION: WITH PEAK, LATEXFREE ELASTIC, HEIGHT 9,5 CM, DIAMETER OF TOP 21 CM MATERIAL: POLYPROPYLENE THICKNESS: PEAK 80G/M, CAP 36G/M, HAIRNET 10-12G/M PACKAGING: 100 PCS/DISPENSERBOX, 1000 PCS/CA 1 * 1.000 Pce
Fournisseur: VWR Collection
SNOOD CAP WITH HAIRNET COLOR: DARK BLUE SIZE: L DESCRIPTION: WITH PEAK, LATEXFREE ELASTIC, HEIGHT 9,5 CM, DIAMETER OF TOP 21 CM MATERIAL: POLYPROPYLENE THICKNESS: PEAK 80G/M, CAP 36G/M, HAIRNET 10-12G/M PACKAGING: 100 PCS/DISPENSERBOX, 1000 PCS/CA 1 * 1.000 Pce
COMMANDE
Développer tout / Réduire tout
SNOOD CAP WITH HAIRNET COLOR: DARK BLUE SIZE: L DESCRIPTION: WITH PEAK, LATEXFREE ELASTIC, HEIGHT 9,5 CM, DIAMETER OF TOP 21 CM MATERIAL: POLYPROPYLENE THICKNESS: PEAK 80G/M², CAP 36G/M², HAIRNET 10-12G/M² PACKAGING: 100 PCS/DISPENSERBOX, 1000 PCS/CA 1 * 1.000 Pce GUNA130466 1 * 1.000 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou