Synonyms:
2-Fluoropropionicacid
H25841.03
H25841.03EA
312
EUR
InStock
H25841.03
2-Fluoropropionic acid 96%
2-Fluoropropionic acid
Purity: 96%
CAS No.: 6087-13-4
Molecular Formula: C3H5FO2
Formula Weight: 92.07
Flash Point: 82°(180°F)
Refractive Index: 1.3830
Odour: Acrid
Boiling Point: 66-67°/30mm
UN No.: UN3265
MDL No.: MFCD06247714