H25961.03
H25961.03EA
142
EUR
InStock
H25961.03
8-Bromoquinoleine ≥98%
8-Bromoquinoleine
Purity: 98%
CAS No.: 16567-18-3
Molecular Formula: C9H6BrN
Formula Weight: 208.06
Flash Point: >110°(230°F)
Refractive Index: 1.6725
Boiling Point: 302-304°
MDL No.: MFCD00191859