Synonyms:
Di-tert-butyl 2,2'-(1,4,7,10-tetraazacyclododecane-1,7-diyl)diacetate, DO2A-tert-butyl ester
H26293.MD
H26293.MDEA
195
EUR
InStock
H26293.MD
1,7-Bis(tert-butoxycarbonylmethyl)-1,4,7,10-tetraazacyclododecane
1,7-Bis(tert-butoxycarbonylmethyl)-1,4,7,10-tetraazacyclododecane
CAS No.: 162148-48-3
Molecular Formula: C20H40N4O4
Formula Weight: 400.56
MDL No.: MFCD09263303