Synonyms:
3,6-Dihydroxy-2-phenyl-4-benzopyrone, 3,6-Dihydroxy-2-phenyl-4H-benzo[b]pyran-4-one, 3,6-Dihydroxy-2-phenyl-4H-chromen-4-one, 3,6-Dihydroxy-2-phenyl-4H-1-benzopyran-4-one, 3,6-Dihydroxy-2-phenylchromen-4-one
H26989.06
H26989.06EA
199
EUR
InStock
H26989.06
3,6-Dihydroxyflavone ≥97%
3,6-Dihydroxyflavone
Purity: 97%
CAS No.: 108238-41-1
Molecular Formula: C15H10O4
Formula Weight: 254.24
Melting Point: 231-234°
MDL No.: MFCD00143082