Synonyms:
Nonafluorobutane-1-sulphonyl chloride
H27437.03
H27437.03EA
86.2
EUR
InStock
H27437.03
Chlorure de perfluorobutane-1-sulfonyle ≥98%
Perfluorobutane-1-sulphonyl chloride
Purity: 98%
CAS No.: 2991-84-6
Molecular Formula: CF3(CF2)3SO2Cl
Formula Weight: 318.55
UN No.: 3265
MDL No.: MFCD03412262