H27644.09
H27644.09EA
169
EUR
InStock
H27644.09
2,4-Dichloro-5-nitropyrimidine ≥97%
2,4-Dichloro-5-nitropyrimidine
Purity: 97%
CAS No.: 49845-33-2
Molecular Formula: C4HCl2N3O2
Formula Weight: 193.98
Flash Point: 110°(230°F)
Melting Point: 28-32°
Boiling Point: 0.9°/0.5mm
MDL No.: MFCD00127867