Synonyms:
Boc-azetidine-3-carboxylic acid, N-Boc-Azetidine-3-carboxylic acid, 1-N-Boc-3-Azetidinecarboxylic acid, 1-Boc-azetidine-3-carboxylic Acid
H28817.03
H28817.03EA
108
EUR
InStock
H28817.03
N-Boc-azetidine-3-carboxylic acid 97%
N-Boc-azetidine-3-carboxylic acid
Purity: 97%
CAS No.: 142253-55-2
Molecular Formula: C9H15NO4
Formula Weight: 201.22
Melting Point: 98-105°
MDL No.: MFCD01860897