H31763.22
H31763.22EA
47.8
EUR
InStock
H31763.22
Acide 2-aminobenzenesulfonique 95%
2-Aminobenzenesulphonic acid
Purity: 97%
CAS No.: 88-21-1
Molecular Formula: C6H7NO3S
Formula Weight: 173.19
Odour: Odourless
Melting Point: 300°
MDL No.: MFCD00007705