Synonyms:
1-(Tetrahydro-2H-pyran-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-imidazole
H32369.ME
H32369.MEEA
566
EUR
InStock
H32369.ME
1-(2-Tetrahydropyranyl)-1H-imidazole-5-boronic acid pinacol ester 95%
1-(2-Tetrahydropyranyl)-1H-imidazole-5-boronic acid pinacol ester
Purity: 95%
CAS No.: 1029684-37-4
Molecular Formula: C14H23BN2O3
Formula Weight: 278.16
Storage Temperature: 2°C to 8°C
Melting Point: 129-133°
MDL No.: MFCD11100960