H32992.03
H32992.03EA
110
EUR
InStock
H32992.03
1H-Imidazol-4-ylacétonitrile 97%
1H-Imidazol-4-ylacétonitrile
Purity: 97%
CAS No.: 18502-05-1
Molecular Formula: C5H5N3
Formula Weight: 107.11
Odour: Odourless
Melting Point: 136-138°
UN No.: UN3439
MDL No.: MFCD00070306