H37600.03
H37600.03EA
106
EUR
InStock
H37600.03
5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalene-2-boronic acid 98%
5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalene-2-boronic acid
Purity: 98%
CAS No.: 169126-63-0
Molecular Formula: C14H21BO2
Formula Weight: 232.13
Physical Form: Powder
Appearance: White
Melting Point: 195-197°
MDL No.: MFCD06801711