H37697.06
H37697.06EA
132
EUR
InStock
H37697.06
Dodécylphosphonate de diéthyle 97+%
Dodécylphosphonate de diéthyle
Purity: 97+%
CAS No.: 4844-38-6
Molecular Formula: C16H35O3P
Formula Weight: 306.42
Physical Form: Liquid
Appearance: Colourless
Boiling Point: 168°/1.5mm
MDL No.: MFCD00015595