H488-68EA
0
EUR
InStock
H488-68
Méthanol ≥99.9% (par GC), UltimAR® pour la chromatographie liquide, pour l'analyse des résidus organiques, pour spectrophotométrie dans l'UV, Macron Fine Chemicals™
Méthanol
UltimAR-solvents that meet ACS specifications for liquid chromatography, UV-spectrophotometry, and general purpose use. They are also tested by GC/ECD and GC/FID for use in extraction/concentration sample clean-up protocols for trace-level organic impurities.
Suitable for liquid chromatography, extract/conc, UV-spectrophotometry, organic residue analysis.