Synonyms:
2,4-Dimethylbenzene-1-sulphonyl chloride, m-Xylene-4-sulphonyl chloride
H50301.03
H50301.03EA
66.1
EUR
InStock
H50301.03
Chlorure de 2,4-diméthylbenzènesulfonyle ≥98%
Chlorure de 2,4-diméthylbenzènesulfonyle
Purity: 98%
CAS No.: 609-60-9
Molecular Formula: C8H9ClO2S
Formula Weight: 204.68
Flash Point: 110°(230°F)
Refractive Index: 1.5550
Odour: Acrid
Melting Point: 32-33°
UN No.: 3261
MDL No.: MFCD00156143