Synonyms:
2,4-Dimethylbenzoylchloride, 2,4-Dimethylbenzene-1-carbonyl chloride
H50387.03
H50387.03EA
199
EUR
InStock
H50387.03
Chlorure de 2,4-diméthylbenzoyle ≥97%
Chlorure de 2,4-diméthylbenzoyle
Purity: 97%
CAS No.: 21900-42-5
Molecular Formula: C9H9ClO
Formula Weight: 168.62
Melting Point: 24-26°
Boiling Point: 82°/1mm
UN No.: 3265
MDL No.: MFCD00798005