Synonyms:
2-(3-Ethynyl-phenyl)-4,4,5,5-tetramethyl-[1,3,2]dioxaborolane, 2-(3-Ethynylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
H51902.03
H51902.03EA
130
EUR
InStock
H51902.03
3-Ethynylbenzeneboronic acid pinacol ester 95%
3-Ethynylbenzeneboronic acid pinacol ester
Purity: 95%
Molecular Formula: C14H17BO2
Formula Weight: 228.10
MDL No.: MFCD16294529