H52311.03
H52311.03EA
287
EUR
InStock
H52311.03
2-Bromo-3-(bromomethyl)pyridine 96%
2-Bromo-3-(bromomethyl)pyridine
Purity: 96%
CAS No.: 94446-97-6
Molecular Formula: C6H5Br2N
Formula Weight: 250.92
Flash Point: 110°(230°F)
Boiling Point: 291°
MDL No.: MFCD08276299