H52722.03
H52722.03EA
92.9
EUR
InStock
H52722.03
3-Chlorocarbonylbenzeneboronic anhydride 90%, tech.
3-Chlorocarbonylbenzeneboronic anhydride
Purity: 90%
CAS No.: 332154-58-2
Molecular Formula: C7H4BClO2
Formula Weight: 166.37
Melting Point: 252-254°
MDL No.: MFCD06801675