Synonyms:
4-n-Propylbenzeneboronic acid, (4-Propylphenyl)boronic acid, 4-n-Propylphenylboronic acid
H52884.06
H52884.06EA
133
EUR
InStock
H52884.06
4-Propylphenylboronic acid ≥98%
4-Propylphenylboronic acid
Purity: 98%
CAS No.: 134150-01-9
Molecular Formula: C9H13BO2
Formula Weight: 164.01
Melting Point: 106-108°
MDL No.: MFCD00859261