Synonyms:
2-Methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-amine, 3-Amino-2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-Methoxy-3-aminopyridine-5-boronic acid pinacol ester, 2-(5-Amino-6-methoxy-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Amino-2-methoxypyridine-5-boronic acid Pinacol ester
H54570.03
H54570.03EA
279
EUR
InStock
H54570.03
3-Amino-2-methoxypyridine-5-boronic acid pinacol ester 96%
3-Amino-2-methoxypyridine-5-boronic acid pinacol ester
Purity: 96%
CAS No.: 893440-50-1
Molecular Formula: C12H19BN2O3
Formula Weight: 250.10
Melting Point: 131-133°
MDL No.: MFCD12923427