H54581.06
H54581.06EA
78.7
EUR
InStock
H54581.06
Benzyloxyacetic acid 95%
Benzyloxyacetic acid
Purity: 95%
CAS No.: 30379-55-6
Molecular Formula: C9H10O3
Formula Weight: 166.17
Flash Point: 113°(235°F)
Refractive Index: 1.526
Boiling Point: 137-139°
MDL No.: MFCD00274211