H55596.06
H55596.06EA
144
EUR
InStock
H55596.06
Tributylphénylétain 97%
Tributylphénylétain
Purity: 97%
CAS No.: 960-16-7
Molecular Formula: C18H32Sn
Formula Weight: 367.14
Flash Point: 113°(235°F)
Physical Form: Liquid
Density: 1.125
Refractive Index: 1.5160
Appearance: Colourless
Boiling Point: 125-128° /0.14mm
Beilstein Reference: 3610571
UN No.: UN2788
MDL No.: MFCD00134394