Stabilization:
Stabilised with sodium carbonate (Na₂CO₃)
H56594.06
H56594.06EA
120
EUR
InStock
H56594.06
(2-Bromoethoxy)-tert-butyldimethylsilane 98% stabilisé
(2-Bromoethoxy)-tert-butyldimethylsilane
Purity: 98%
CAS No.: 86864-60-0
Molecular Formula: C8H19BrOSi
Formula Weight: 239.23
Flash Point: 73°(163°F)
Physical Form: Liquid
Density: 1.115
Refractive Index: 1.444
Appearance: Colourless
Boiling Point: 120°/100mm
MDL No.: MFCD00209550