H56994.06
H56994.06EA
98.8
EUR
InStock
H56994.06
5-Norbornene-2-ol (mélange endo + exo) 99%
5-Norbornene-2-ol
Purity: 99%
CAS No.: 13080-90-5
Molecular Formula: C7H10O
Formula Weight: 110.15
Flash Point: 51°(124°F)
Physical Form: Solid
Melting Point: 104-106°
UN No.: UN1325
MDL No.: MFCD00167566