Synonyms:
(S)-1-tert-Butyl 2-ethyl 4-oxopiperidine-1,2-dicarboxylate, Ethyl (S)-1-Boc-4-oxopiperidine-2-carboxylate
H57213.03
H57213.03EA
166
EUR
InStock
H57213.03
(S)-1-Boc-4-oxo-piperidine-2-carboxylic acid methyl ester 95%
(S)-1-Boc-4-oxo-piperidine-2-carboxylic acid methyl ester
Purity: 95%
CAS No.: 180854-44-8
Molecular Formula: C13H21NO5
Formula Weight: 271.31
Storage Temperature: 2°C to 8°C
Flash Point: 110°(230°F)
Refractive Index: 1.4600
Optical Rotation: -20° (c=0.5 in chloroform)
MDL No.: MFCD15071881