Synonyms:
(2S)-4-Oxopiperidine-2-carboxylic acid, N-BOC protected, 4-Oxo-L-pipecolinic acid, N-BOC protected, (S)-(-)-1-Boc-4-oxopiperidine-2-carboxylic acid, (S)-1-Boc-4-oxopipecolic acid, (2S)-1-(tert-Butoxycarbonyl)-4-oxopiperidine-2-carboxylic acid
H57617.03
H57617.03EA
120
EUR
InStock
H57617.03
(S)-1-(tert-Butoxycarbonyl)-4-oxopiperidine-2-carboxylic acid 95%
(S)-1-(tert-Butoxycarbonyl)-4-oxopiperidine-2-carboxylic acid
Purity: 95%
CAS No.: 198646-60-5
Molecular Formula: C11H17NO5
Formula Weight: 243.26
Melting Point: 121-126°
Optical Rotation: -22° (c=1 in chloroform)
MDL No.: MFCD03094867