Packaged under argon in resealable ChemSeal™ bottles.
H58167.AD
H58167.ADEA
208
EUR
InStock
H58167.AD
3,4-Difluorobenzylzinc bromide 0.5 M dans le THF
3,4-Difluorobenzylzinc bromide
Packaged under Argon in resealable ChemSeal™ bottles
CAS No.: 307496-34-0
Molecular Formula: C7H5BrF2Zn
Formula Weight: 272.40
Storage Temperature: 2°C to 8°C
UN No.: UN3399
MDL No.: MFCD01311420